EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Highly active dizinc catalyst for the copolymerization of carbon dioxide and cyclohexene oxide at one atmosphere pressure.
PMID 19115338
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C102504
Cyclohexene oxide, 98%
C6H10O