EMAIL THIS PAGE TO A FRIEND
Endocrinology
Sphingosine kinase as an oncogene: autocrine sphingosine 1-phosphate modulates ML-1 thyroid carcinoma cell migration by a mechanism dependent on protein kinase C-alpha and ERK1/2.
PMID 19116345
Abstract
Sphingosine 1-phosphate (S1P) induces migration of the human thyroid follicular carcinoma cell line ML-1 by activation of S1P(1) and S1P(3) receptors, G(i) proteins, and the phosphatidylinositol 3-kinase-Akt pathway. Because sphingosine kinase isoform 1 (SK) recently has been implicated as an oncogene in various cancer cell systems, we investigated the functions of SK in the migration, proliferation and adhesion of the ML-1 cell line. SK overexpressing ML-1 cells show an enhanced secretion of S1P, which can be attenuated, by inhibiting SK activity and a multidrug-resistant transport protein (ATP-binding cassette transporter). Furthermore, overexpression of SK enhances serum-induced migration of ML-1 cells, which can be attenuated by blocking ATP-binding cassette transporter and SK, suggesting that the migration is mediated by autocrine signaling through secretion of S1P. Inhibition of protein kinase C alpha, with both small interfering RNA (siRNA) and small molecular inhibitors attenuates migration in SK overexpressing cells. In addition, SK-overexpressing cells show an impaired adhesion, slower cell growth, and an up-regulation of ERK1/2 phosphorylation, as compared with cells expressing a dominant-negative SK. Taken together, we present evidence suggesting that SK enhances migration of ML-1 cells by an autocrine mechanism and that the S1P-evoked migration is dependent on protein kinase C alpha, ERK1/2, and SK.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
ML-1, 88113007