EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Nitration of cathepsin D enhances its proteolytic activity during mammary gland remodelling after lactation.
PMID 19125694
Abstract
Proteomic studies in the mammary gland of control lactating and weaned rats have shown that there is an increased pattern of nitrated proteins during weaning when compared with controls. Here we report the novel finding that cathepsin D is nitrated during weaning. The expression and protein levels of this enzyme are increased after 8 h of litter removal and this up-regulation declines 5 days after weaning. However, there is a marked delay in cathepsin D activity since it does not increase until 2 days post-weaning and remains high thereafter. In order to find out whether nitration of cathepsin D regulates its activity, iNOS (inducible nitric oxide synthase)(-/-) mice were used. The expression and protein levels of this enzyme were similar to WT (wild-type) animals, but the proteolytic activity was significantly reduced during weaning in knockout compared to WT mice. in vitro treatment of recombinant human cathepsin D or lactating mammary gland homogenates with relatively low concentrations of peroxynitrite enhances the nitration as well as specific activity of this enzyme. Using MS, it has been shown that the residue Tyr168 was nitrated. All of these results show that protein nitration during weaning might be a signalling pathway involved in mammary gland remodelling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2032 Pepstatin A−Agarose, saline suspension