EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Production of stilbenoids and phenolic acids by the peanut plant at early stages of growth.
PMID 19127717
Abstract
The peanut plant (Arachis hypogaea) is known to produce stilbene phytoalexins as a defensive response to fungal invasion; however, the distribution of phytoalexins among different organs of the peanut plant at early stages of growth under axenic conditions has not been studied. Axenic plants produced a stilbenoid, resveratrol, as well as soluble bound and free phenolic acids, including 4-methoxycinnamic acid, which is reported in peanuts for the first time. Neither resveratrol nor phenolic acids were found in the root mucilage; the prenylated stilbenes were restricted to the mucilage and were not found in other organs of the peanut plant. These findings may lead to a better understanding of the defensive role of peanut stilbenes and phenolic acids.
Related Materials