EMAIL THIS PAGE TO A FRIEND
Journal of clinical hypertension (Greenwich, Conn.)
Gingival hyperplasia in a patient with hypertension.
PMID 19128276
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0165
Nisoldipine, ≥98% (HPLC)
C20H24N2O6