EMAIL THIS PAGE TO A FRIEND
Mycopathologia
Oral potassium iodide for the treatment of sporotrichosis.
PMID 19130293
Abstract
Potassium Iodide is the antimycotic of choice for the treatment of cutaneous sporotrichosis, because of its efficacy, safety and low cost. We carried out a review of published studies on the benefits and adverse reactions of using SSKI (Saturated Solution Potassium Iodide) as treatment for sporotrichosis, but could not identify any well-designed clinical trails. There is an urgent need to conduct randomized double-blind placebo-controlled trials and critically assess usefulness of SSKI by using a standardize monitoring or an effective self-report system.