EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Acute toxic effects of three pesticides on Pseudomonas putida monitored by microcalorimeter.
PMID 19130374
Abstract
A series of calorimetric experiments were performed to investigate the toxic effects of beta-cypermethrin (BCP), bensulfuron-methyl (BSM) and prometryne (PM) on Pseudomonas putida (P. putida). The metabolic action of P. putida on the three pesticides was studied by obtaining power-time curves. The growth of P. putida was inhibited completely in each case when the concentrations of pesticides were up to 80 micro g mL(- 1). The relationships between the inhibitory ratio (k) and doses of contaminants were approximately linear for the three pesticides. The total heat dissipated per milliliter (Q(total)) for the pesticides decreased during the course of the experiment. The OD(600) of P. putida growth in the absence and presence of pesticides was also obtained. The power-time curves of P. putida growth coincided with its turbidity curves. This elucidates that microcalorimetric method agrees well with the routine microbiological method. Among these three pesticides, BSM was found to be the most toxic with an IC(50) of 19.24 micro g mL(- 1) against P. putida. PM exhibited moderate virulence with an IC(50) of 27.86 micro g mL(- 1) and BCP had the lowest toxicity with an IC(50) of 39.64 micro g mL(- 1).
Related Materials