EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
The chemistry of escapin: identification and quantification of the components in the complex mixture generated by an L-amino acid oxidase in the defensive secretion of the sea snail Aplysia californica.
PMID 19130530
Abstract
Escapin is an L-amino acid oxidase in the ink of a marine snail, the sea hare Aplysia californica, which oxidizes L-lysine (1) to produce a mixture of chemicals which is antipredatory and antimicrobial. The goal of our study was to determine the identity and relative abundance of the constituents of this mixture, using molecules generated enzymatically with escapin and also using products of organic syntheses. We examined this mixture under the natural range of pH values for ink-from approximately 5 at full strength to approximately 8 when fully diluted in sea water. The enzymatic reaction likely forms an equilibrium mixture containing the linear form alpha-keto-epsilon-aminocaproic acid (2), the cyclic imine Delta(1)-piperidine-2-carboxylic acid (3), the cyclic enamine Delta(2)-piperidine-2-carboxylic acid (4), possibly the linear enol 6-amino-2-hydroxy-hex-2-enoic acid (7), the alpha-dihydroxy acid 6-amino-2,2-dihydroxy-hexanoic acid (8), and the cyclic aminol 2-hydroxy-piperidine-2-carboxylic acid (9). Using NMR and mass spectroscopy, we show that 3 is the major component of this enzymatic product at any pH, but at more basic conditions, the equilibrium shifts to produce relatively more 4, and at acidic conditions, the equilibrium shifts to produce relatively more 2, 7, and/or 9. Studies of escapin's enzyme kinetics demonstrate that because of the high concentrations of escapin and L-lysine in the ink secretion, millimolar concentrations of 3, H(2)O(2), and ammonia are produced, and also lower concentrations of 2, 4, 7, and 9 as a result. We also show that reactions of this mixture with H(2)O(2) produce delta-aminovaleric acid (5) and delta-valerolactam (6), with 6 being the dominant component under the naturally acidic conditions of ink. Thus, the product of escapin's action on L-lysine contains an equilibrium mixture that is more complex than previously known for any L-amino acid oxidase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123188
5-Aminovaleric acid, 97%
C5H11NO2
194336
5-Aminovaleric acid hydrochloride, 99%
C5H11NO2 · HCl
V209
δ-Valerolactam, 98%
C5H9NO