EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Cancer preventive agents. Part 8: Chemopreventive effects of stevioside and related compounds.
PMID 19131254
Abstract
In a search for potential cancer chemopreventive agents from natural resources, stevioside (1), a sweetener, and six related compounds, including two aglycones steviol (6) and isosteviol (7), were screened in an in vitro assay for inhibitory effects on Epstein-Barr virus early antigen activation. Compounds 1, 6 and 7 showed significant activity in this assay and also exhibited strong inhibitory effects in a two-stage carcinogenesis test using mouse skin induced by 7,12-dimethylbenz[a]anthracene (DMBA) and 12-O-tetradecanoylphorbol-13-acetate (TPA). The inhibitory effects of these three compounds were greater than that of glycyrrhizin. Furthermore, these three compounds significantly inhibited mouse skin carcinogenesis initiated by peroxynitrite and promoted by TPA. Their activities were comparable to that of curcumin. These results suggested that 1, as well as 6 and 7, could be valuable as chemopreventive agents for chemical carcinogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19345
Steviol, analytical standard
C20H30O3
H8664
Steviol hydrate, ≥98% (HPLC), powder
C20H30O3 · xH2O