EMAIL THIS PAGE TO A FRIEND
Evidence-based complementary and alternative medicine : eCAM
A chinese herbal decoction, danggui buxue tang, stimulates proliferation, differentiation and gene expression of cultured osteosarcoma cells: genomic approach to reveal specific gene activation.
PMID 19131392
Abstract
Danggui Buxue Tang (DBT), a Chinese herbal decoction used to treat ailments in women, contains Radix Astragali (Huangqi; RA) and Radix Angelicae Sinensis (Danggui; RAS). When DBT was applied onto cultured MG-63 cells, an increase of cell proliferation and differentiation of MG-63 cell were revealed: both of these effects were significantly higher in DBT than RA or RAS extract. To search for the biological markers that are specifically regulated by DBT, DNA microarray was used to reveal the gene expression profiling of DBT in MG-63 cells as compared to that of RA- or RAS-treated cells. Amongst 883 DBT-regulated genes, 403 of them are specifically regulated by DBT treatment, including CCL-2, CCL-7, CCL-8, and galectin-9. The signaling cascade of this DBT-regulated gene expression was also elucidated in cultured MG-63 cells. The current results reveal the potential usage of this herbal decoction in treating osteoporosis and suggest the uniqueness of Chinese herbal decoction that requires a well-defined formulation. The DBT-regulated genes in the culture could serve as biological responsive markers for quality assurance of the herbal preparation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6178
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
F4135
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
F0392
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
12107C
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN® process, sterile-filtered, suitable for cell culture
12303C
Fetal Bovine Serum, USDA approved countries, sterile-filtered, cell culture tested
F1051
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
12003C
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
12007C
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
12203C
Fetal Bovine Serum, New Zealand origin, sterile-filtered, suitable for cell culture
F0926
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
12306C
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
12006C
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
12207C
Fetal Bovine Serum, New Zealand origin, cell culture tested
F9423
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
F3297
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
12105C
Fetal Bovine Serum, Dialyzed by ultrafiltration against 0.15M NaCl, USA origin, sterile-filtered, cell culture tested
12103C
Fetal Bovine Serum, USA origin, cell culture tested
12106C
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
F6765
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
F7305
Fetal Bovine Serum, Mouse Embryonic Stem Cell Screened, suitable for cell culture, sterile-filtered, USDA Approved