EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Dual organocatalysis: asymmetric allylic-allylic alkylation of alpha,alpha-dicyanoalkenes and Morita-Baylis-Hillman carbonates.
PMID 19132709
Abstract
The first highly enantioselective allylic-allylic alkylation of alpha,alpha-dicyanoalkenes and Morita-Baylis-Hillman carbonates by dual catalysis of (DHQD)(2)AQN and (S)-BINOL has been investigated. Excellent stereoselectivities have been achieved for a broad spectrum of substrates (d.r. > 99:1, up to 99 % ee). The multifunctional allylic products could be efficiently converted to a range of complex chiral cyclic frameworks. EWG = electron-withdrawing group, (DHQD)(2)AQN = hydroquinidine (anthraquinone-1,4-diyl) diether, (S)-BINOL = (S)-(-)-1,1'-bi-2-naphthol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104655
1,1′-Bi-2-naphthol, 99%
C20H14O2