EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Simultaneous spectrophotometric determination of binary mixtures of surfactants using continuous wavelet transformation.
PMID 19135304
Abstract
This work presents a simple, rapid, and novel method for simultaneous determination of binary mixtures of some surfactants using continuous wavelet transformation. The method is based on the difference in the effect of surfactants Cetyltrimethylammoniumbromide (CTAB), dodecyl trimethylammonium bromide (DTAB), cetylpyridinium bromide (CPB) and TritonX-100 (TX-100) on the absorption spectra of complex of Beryllium with Chrome Azurol S (CAS) at pH 5.4. Binary mixtures of CTAB-DTAB, DTAB-CPB and CTAB-TX-100 were analyzed without prior separation steps. Different mother wavelets from the family of continuous wavelet transforms were selected and applied under the optimal conditions for simultaneous determinations. The proposed methods, under the working conditions, were successfully applied to simultaneous determination of surfactants in hair conditioner and mouthwash samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199532
Chromeazurol S, Dye content 50 %
C23H13Cl2Na3O9S
30725
Decyltrimethylammonium bromide, ≥98.0% (NT)
C13H30BrN