EMAIL THIS PAGE TO A FRIEND
Oncogene
Specific regulation of mRNA cap methylation by the c-Myc and E2F1 transcription factors.
PMID 19137018
Abstract
Methylation of the mRNA 5' guanosine cap is essential for efficient gene expression. The 5' methyl cap binds to eIF4E, which is the first step in the recruitment of mRNA to the 40S ribosomal subunit. To investigate whether mRNA cap methylation is regulated in a gene-specific manner, we established a method to detect the relative level of cap methylation on specific mRNAs. We found that two transcription factors, c-Myc and E2F1, induce cap methylation of their transcriptional target genes, and therefore, c-Myc and E2F1 upregulate gene expression by simultaneously inducing transcription and promoting translation. c-Myc-induced cap methylation is greater than transcriptional induction for the majority of its target genes, indicating that this is a major mechanism by which Myc regulates gene expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6133
7-Methylguanosine 5′-triphosphate sodium salt, ≥85% (HPLC)
C11H18N5O14P3 · xNa+