EMAIL THIS PAGE TO A FRIEND
Molecular Cancer Therapeutics
Gemcitabine sensitization by checkpoint kinase 1 inhibition correlates with inhibition of a Rad51 DNA damage response in pancreatic cancer cells.
PMID 19139112