EMAIL THIS PAGE TO A FRIEND
Toxicology and industrial health
Deleterious effects induced by oxidative stress in liver nuclei from rats receiving an alcohol-containing liquid diet.
PMID 19141567
Abstract
Highly purified rat-liver nuclei were previously shown to have nuclear ethanol (EtOH) metabolizing system able to bioactivate alcohol to acetaldehyde and 1-hydroxyethyl radicals. These reactive metabolites were able to covalently bind to nuclear proteins and lipids potentially being able to provoke oxidative stress of nuclear components. In this study, the above-mentioned possibility was explored. Sprague Dawley male rats (125-150 g) were fed a standard Lieber and De Carli liquid diet for 28 days. Controls were pair-fed with a diet, in which EtOH was isocalorically replaced with carbohydrate. The presence of a chlorzoxazone hydroxylase activity inducible by the repetitive EtOH drinking further suggested the presence of CYP2E1 in the highly purified nuclei. Nuclei from EtOH-drinking rats evidenced significantly increased susceptibility to a t-butyl hydroperoxide challenge as detected by chemiluminescence emission, increased formation of protein carbonyls, and decreased content of protein sulfhydryls. In contrast, no significant changes in the nuclear lipid hydroperoxides formation or even decreases in the 8-oxo-7,8-dihydro-2-deoxyguanosine were observed. No significant differences were observed in different parameters of the alkaline Comet assay. In immunohistochemical studies performed, no expression of p53 was observed in the livers of the animals under the experimental conditions tested. Since nuclear proteins and lipids are known to play a role in cell growth, differentiation, repair and signaling, their alterations by either oxidative stress, or by covalent binding might be of relevance to liver tumor promotion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC148
6-Hydroxychlorzoxazone, ≥98% (HPLC)
C7H4ClNO3
C188
6-Hydroxychlorzoxazone, solid
C7H4ClNO3