EMAIL THIS PAGE TO A FRIEND
Roczniki Panstwowego Zakladu Higieny
[Occurrence of plant protection product residues in apples in 2007].
PMID 19143426
Abstract
The aim of this paper was to present occurrence of plant protection product residues in apples in 2007. Gas chromatographic and spectroscopy methods were used. The most frequently found were fungicides: dithiocarbamate residues (18% of the analysed samples), captan residues (13%), pyrimethanil residues (6%) and insecticides: chlorpiryfos residues (6%), diazinon residues (6%) and pirimicarb residues (5%). 5.6% of analysed samples exceeded the national Maximum Residue Level established for apples. Violations were found for: flusilazole (n = 2), propiconazole (n = 2), cyprodinil (n = 1), dimethoate (n = 1), fenitrothion (n = 1).