EMAIL THIS PAGE TO A FRIEND
Diabetes self-management
Anti-inflammatory diets. Hype or hope?
PMID 19145758
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0070
Salsalate, ≥98% (HPLC)
C14H10O5