EMAIL THIS PAGE TO A FRIEND
The International journal of periodontics & restorative dentistry
Hard and soft tissue responses to the platform-switching technique.
PMID 19146050
Abstract
Platform switching is a concept recently introduced in implant dentistry. It is intended to reduce the crestal bone loss that is commonly found around implants exposed to the oral environment. The aim of this study was to examine biopsy specimens to help explain the biologic processes occurring around a platform-switched implant. A mandibular implant was removed 2 months after placement because of prosthetic rehabilitation difficulties. The implant was then sectioned and subjected to histologic and histomorphometric analysis. An inflammatory connective tissue infiltrate was localized over the entire surface of the implant platform and approximately 0.35 mm coronal to the implant-abutment junction, along the healing abutment. A possible reason for bone preservation around a platform-switched implant may lie in the inward shift of the inflammatory connective tissue zone at the implant-abutment junction, which reduces its injurious effect on the alveolar bone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8129
Acid Fuchsin, used in tissue staining
C20H17N3Na2O9S3