EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on glycosides from Gentiana veitchiorum].
PMID 19149260
Abstract
To study the chemical constituents of Gentiana veitchiorum. The chemical constituents were isolated by chromatography and identified by spectral data. Five glycosides, loganic acid (1), gentiopicroside (2), isoorientin 3'-methyl ether (3), isovitexin (4), isoorientin (5) were isolated and identified. Compounds 1-5 were isolated from this plant for the first time.