EMAIL THIS PAGE TO A FRIEND
Chemosphere
An improved method for assessing relative nucleophilic substitution reactivities of polychlorinated benzenes.
PMID 19152956
Abstract
An existing substitution reaction method was improved as part of a system to measure the persistency of selected chemicals in the environment by evaluating their chemical reactivity. Hexachlorobenzene (HCB), pentachlorobenzene, 1,2,4,5-tetrachlorobenzene and 1,2,4-trichlorobenzene were selected as model compounds. Sodium methoxide in methanol was used as a nucleophile and found to be stable for at least 35 days after preparation. The substitution reaction system was modified so that nitrogen protection was not necessary to avoid oxidation and hydrolysis effects, which led to improved results. The reactivity of polychlorinated benzenes (PCBz), which substituted chlorine for methoxide according to a second-order rate constant, increased as the number of chlorine atoms on the benzene ring increased. HCB was selected as a standard for the k(2) calculations of the substitution reactions. A normalized k(2) (k(N)) was calculated as k(N)=k(PCBz)/k(HCB). GC-MS analysis confirmed that the reactions were pure nucleophilic aromatic substitutions without side reactions.