Molecular immunology
Toll-like receptor 7 cooperates with IL-4 in activated B cells through antigen receptor or CD38 and induces class switch recombination and IgG1 production.
PMID 19157556
IL-4 and 8-mercaptoguanosine (8-SGuo) stimulation of CD38-activated B cells induces mu to gamma1 class switch recombination (CSR) at the DNA level leading to a high level of IgG1 production. Although some of signaling events initiated by IL-4 in activated B cells have been characterized, the involvement of TLR/MyD88 and Btk pathway in IL-4-dependent mu to gamma1 CSR has not been thoroughly evaluated. In this study, we characterized receptors for 8-SGuo and differential roles of 8-SGuo and IL-4 in the induction and mu to gamma1 CSR and IgG1 production. The role of TLR7 and MyD88 in 8-SGuo-induced AID expression and mu to gamma1 CSR was documented, as 8-SGuo did not act on CD38-stimulated splenic B cells from Tlr7(-/-) and Myd88(-/-) mice. CD38-activated B cells from Btk-deficient mice failed to respond to TLR7 ligands for the AID expression and CSR, indicating that Btk is also indispensable for the system. Stimulation of CD38-activated B cells with 8-SGuo induced significant AID expression and DNA double strand breaks, but IL-4 stimulation by itself did not trigger mu to gamma1 CSR. Intriguingly, the mu to gamma1 CSR in the B cells stimulated with CD38 and 8-SGuo totally depends on IL-4 stimulation. Similar results were obtained in the activated B cells through BCR and loxoribine, a well-known TLR7 ligand, in place of 8-SGuo. In vivo administration of TLR7 ligand and anti-CD38 antibody induced the generation of CD138(+) IgG1(+) cells. These results indicate that TLR7 is a receptor for 8-SGuo and plays an essential role in the AID and Blimp-1 expression; however it is not enough to complete mu to gamma1 CSR in CD38-activated B cells. IL-4 may be required for the induction of DNA repair system together with AID for the completion of CSR.
Related Materials
Product #
Molecular Formula
Add to Cart
7-Allyl-7,8-dihydro-8-oxoguanosine, 95%