EMAIL THIS PAGE TO A FRIEND
Journal of proteome research
Glycoproteomics analysis of human liver tissue by combination of multiple enzyme digestion and hydrazide chemistry.
PMID 19159218
Abstract
The study of protein glycosylation has lagged far behind the progress of current proteomics because of the enormous complexity, wide dynamic range distribution and low stoichiometric modification of glycoprotein. Solid phase extraction of tryptic N-glycopeptides by hydrazide chemistry is becoming a popular protocol for the analysis of N-glycoproteome. However, in silico digestion of proteins in human proteome database by trypsin indicates that a significant percentage of tryptic N-glycopeptides is not in the preferred detection mass range of shotgun proteomics approach, that is, from 800 to 3500 Da. And the quite big size of glycan groups may block trypsin to access the K, R residues near N-glycosites for digestion, which will result in generation of big glycopeptides. Thus many N-glycosites could not be localized if only trypsin was used to digest proteins. Herein, we describe a comprehensive way to analyze the N-glycoproteome of human liver tissue by combination of hydrazide chemistry method and multiple enzyme digestion. The lysate of human liver tissue was digested with three proteases, that is, trypsin, pepsin and thermolysin, with different specificities, separately. Use of trypsin alone resulted in identification of 622 N-glycosites, while using pepsin and thermolysin resulted in identification of 317 additional N-glycosites. Among the 317 additional N-glycosites, 98 (30.9%) could not be identified by trypsin in theory because the corresponding in silico tryptic peptides are either too small or too big to detect in mass spectrometer. This study clearly demonstrated that the coverage of N-glycosites could be significantly increased due to the adoption of multiple enzyme digestion. A total number of 939 N-glycosites were identified confidently, covering 523 noredundant glycoproteins from human liver tissue, which leads to the establishment of the largest data set of glycoproteome from human liver up to now.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1665 5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg
G5160 β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid
G1875 β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
G4142 β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
G4155 β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
G5635 β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
G6008 β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, 250-600 units/mg protein
G3153 β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
48275 β-Galactosidase from Escherichia coli, for enzyme immunoassay, lyophilized, powder, ~140 U/mg
G8511 β-Galactosidase from Escherichia coli, For use as a marker in SDS-PAGE
G5758 Glucose-6-Phosphatase from rabbit liver, ≥0.05 units/mg protein, Protein ~30 % (balance mostly sucrose)
A9253 L-Amino Acid Oxidase from Crotalus adamanteus, Type I, dried venom, ≥0.3 unit/mg solid
A9378 L-Amino Acid Oxidase from Crotalus adamanteus, Type IV, ≥3.0 units/mg protein (biuret), aqueous suspension
A5147 L-Amino Acid Oxidase from Crotalus atrox (Western Diamondback Rattlesnake), Type VI, dried venom, ≥0.1 unit/mg solid
62287 Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
L4777
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
L4277 Lipase from Aspergillus oryzae, ≥20,000 U/g
62285 Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
02569 Lipase from Candida antarctica, lyophilized, powder, beige, ~0.3 U/mg
62302 Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
62316 Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
L3170
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
L8906 Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
L9031 Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
62298 Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
L0382 Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
62309 Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
L9518
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
62310 Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg