EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Study on safe use of 10% difenoconazole in GAP of Gentiana scabra].
PMID 19160793
Abstract
To investigate the safe use of 10% difenoconazole in planting Gentiana scabra. The degradation dynamics of 10% ifenoconazole in the stems and leaves of G. scabra collecting in different time were determined by GC with ECD detection, and the half life of difenoconazole in the plant was calculated, and then the safe use method of 10% difenoconazole was formulated. Under the local climatic conditions, the half life of 10% difenoconazole was 6.84-6.90 days. In the good agricultural practice (GAP) of G. scabra, the maximal concentration of 10% difenoconazole is 400 g x ha(-1), the safety interval of using 10% difenoconazole is 40 days.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36531
Difenoconazol, PESTANAL®, analytical standard
C19H17Cl2N3O3