EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Mononuclear ferrocenophane structural motifs with two thiourea arms acting as a dual binding site for anions and cations.
PMID 19161309
Abstract
The synthesis of a new type of mononuclear ferrocenophane-based thiourea, in which the ferrocene moiety is simultaneously attached to two thiourea groups directly from 1,1'-bis(isothiocyanato)ferrocene, is reported. These nitrogen-rich structural motifs show remarkable ion-sensing properties because of the presence of the redox active ferrocene unit and the thiourea bridges, which unexpectedly act as a dual binding site for anions and metal ions. They display a selective downfield shift of the thiourea protons and a remarkable cathodic shift of the ferrocene/ferrocenium redox couple with F(-), AcO(-), H(2)PO(4)(-), and HP(2)O(7)(3-) anions, whereas the selective recognition of Hg(2+) metal cations is achieved either by electrochemical or by spectral measurements. The preferred binding modes are proposed for the most representative complexes by means of density functional theory based theoretical calculations showing the Janus-like faces of the receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388297
Ferrocenium hexafluorophosphate, 97%
C10H10F6FeP
482358
Ferrocenium tetrafluoroborate, technical grade
C10H10BF4Fe