EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
A novel class of allosteric modulators of AMPA/Kainate receptors.
PMID 19162477
Abstract
The rapid hydrolysis in vivo of IDRA21 to 2-amino-5-chlorobenzensulfonamide has been demonstrated by microdialysis experiments. The IDRA21 metabolite possess in vitro a biological activity similar to that of IDRA21 itself. Taking 2-amino-5-chlorobenzensulfonamide as lead compound, a novel class of AMPAR positive allosteric modulators has been prepared.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5773
IDRA 21, ≥98%
C8H9ClN2O2S