EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Concentrating and determining the levels of microquantities of bancol in the air and water].
PMID 1916352
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33863
Bensultap, PESTANAL®, analytical standard
C17H21NO4S4