EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Evaluation of changes in memory and learning processes caused by pesticide poisoning].
PMID 1916374
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32538
Pyridaphenthion, PESTANAL®, analytical standard
C14H17N2O4PS