EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on chemical constituents of aerial parts of Ammopiptanthus mongolicus].
PMID 19166007
Abstract
To investigate the chemical constituents of the aerial parts of Ammopiptanthus mongolicus. The chemical constituents were isolated by various column chromatographic methods. The structures were identified by spectral data. Ten compounds were isolated and identified as m-hydroxybenzoic acid (1), 1-(4-hydroxyphenyl) ethanone (2), beta-sitosterol (3), (-)-syringaresinol (4), (+)-lariciresinol (5), blumenol A (6), blumenol B (7), beta-daucosterol (8), coniferin (9), syringin (10). The ten compounds were obtained from the genus Ammopiptanthus for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00103
Abietin, ≥90% (LC/MS-ELSD)
C16H22O8
90974
Eleutheroside B, analytical standard
C17H24O9