EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
Urinary excretion of lithocholic acid and its conjugates by the bile duct-ligated rat.
PMID 1916672
Abstract
The 3-O-glucuronide of lithocholic acid has been shown to be a potent cholestatic agent in rats. However, even after the onset of lithocholic acid glucuronide-induced cholestasis, little of the administered material was recovered in urine. To determine whether this phenomenon was related to the steroid moiety or the form of conjugation, small doses of radiolabeled lithocholic acid glucuronide, lithocholic acid, taurolithocholic acid and/or lithocholic acid sulfate were administered to rats with ligated bile ducts. Urinary excretion of isotope was followed for 24 hr and urinary metabolites of the administered compounds were identified by thin-layer chromatography. Lithocholic and taurolithocholic acids were slowly but relatively efficiently excreted in urine with 73% and 91% of the dose, respectively, recovered in urine over 24 hr. More than 80% of the label in urine from animals receiving these two compounds was in the form of taurine-conjugated beta-muricholic acid. In contrast, lithocholic acid 3-glucuronide and 3-sulfate were poorly excreted: 9% and 12% of the administered doses, respectively, were recovered in urine in 24 hr. Of the small amount of label in urine from rats given the glucuronide, 90% was identified as lithocholic and taurolithocholic acid glucuronides. When lithocholic acid sulfate was given, thin-layer chromatography of urine showed two peaks, which were tentatively identified as tauromurideoxycholic and taurolithocholic acid sulfates. More definitive identification was not possible because of the small amount of the administered dose excreted in urine in these forms.(ABSTRACT TRUNCATED AT 250 WORDS)