EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology
The effects of 2-bromopalmitate on the fatty acid composition in differentiating adipocytes of red sea bream (Pagrus major).
PMID 19166960
Abstract
To determine whether external factors affect the adipogenic function of fish adipocytes, the effects of 2-bromopalmitate (a PPAR agonist) on the fatty acid composition in differentiating adipocytes of red sea bream were investigated in vitro. In the presence of 2-bromopalmitate, the red sea bream adipocytes were differentiated and the effects on the fatty acid composition and the adipogenic gene expression were analyzed. With the level of 2-bromopalmitate, the content of 16:1n-7, a delta-9 desaturation product, increased in association with the increase in a stearoyl CoA desaturase (SCD) gene expression level while the triglyceride accumulation was not affected. Subsequently, the effects on the bioconversion of the n-3 and n-6 fatty acids, which are main series of dietary essential fatty acids, were examined. In the presence of 300 microM of 18:3n-3 or 18:2n-6, red sea bream stromal-vascular cells accumulated the lipid in the cytoplasm within 3 days by the fatty acid uptake with the increase of corresponding fatty acid contents. Furthermore, in both the 18:3n-3 and 18:2n-6 stored cells, the products of delta-6 desaturation (18:4n-3 and 18:3n-6, respectively) and C(18-20) elongation (20:3n-3 and 20:2n-6, respectively) were detected. However, neither the delta-6 desatutration nor C(18-20) elongation of 18:3n-3 and 18:2n-6 were enhanced by 2-bromopalmitate treatment. In conclusion, the results indicate that the adipocyte function in fish, e.g. adipogenic gene expression and fatty acid composition, can be modified by external factors and a main effect of 2-bromopalmitate is the increase in the content of delta-9 desaturation product by stimulating the SCD gene expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21604
2-Bromohexadecanoic acid, ≥99.0% (GC)
C16H31BrO2
238422
2-Bromohexadecanoic acid, ~97%
C16H31BrO2