EMAIL THIS PAGE TO A FRIEND
International angiology : a journal of the International Union of Angiology
Buflomedil: potential new indications for an old agent.
PMID 19174750
Abstract
Our understanding of vascular pathophysiology has significantly improved during the past two decades. Patients with type 2 diabetes mellitus have an increased vascular risk and a series of modifiable risk factors play a crucial role in the atherosclerotic process. The microvascular dysfunction in diabetes results in increased vascular permeability and impaired regulation of blood flow and vascular tone. These changes culminate in nephropathy, retinopathy and neuropathy and probably contribute to the increase vascular morbidity and mortality in this population. Moreover, studies in the skin microvasculature suggest that this microvascular dysfunction contributes significantly to the pathogenesis of diabetic foot. Several studies showed a beneficial effect of vasoactive drugs, including buflomedil, in non-diabetic patients. However, it remains to be established whether these drugs could also be beneficial in the diabetic population, especially in the early stages of diabetes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5899
Buflomedil hydrochloride, analytical standard
C17H25NO4 · HCl