EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Ca2+ pathway involved in the refilling of store sites in rat adrenal medullary cells.
PMID 19176761
Abstract
It has been suggested that store-operated Ca(2+) entry (SOC) facilitates catecholamine secretion and synthesis in bovine adrenal medullary (AM) cells. However, there has been no experimental result clearly showing that cation channel activity is enhanced by store Ca(2+) depletion. Thus the present experiments were undertaken to address the issue of whether rat AM cells have SOC channels. Inhibition of the sarco(endo)plasmic reticulum Ca(2+) (SERCA) pump resulted in a sustained increase in intracellular Ca(2+) concentration ([Ca(2+)](i)) in rat AM cells. This increase was completely suppressed by 2 mM Ni(2+) but not by 100 muM D600. A bath application of Ni(2+), but not D600, produced an outward current at -60 mV in rat AM cells, whereas exposure to a SERCA pump inhibitor did not affect either the whole cell current level or the Ni(2+)-induced outward current. The refilling of intracellular store sites was suppressed by the addition of Ni(2+) to the perfusate. RT-PCR revealed that transcripts for transient receptor potential channels 1 (TRPC1) and 5 (TRPC5) were present in rat adrenal medullas. Immunocytochemistry showed that TRPC1 channels, which have been implicated in SOC in certain types of cells, were mainly localized in the endoplasmic reticulum (ER) and not in the plasma membrane, and that STIM1, a Ca(2+) sensor in the ER, was not expressed in rat AM cells. On the basis of these results, we conclude that rat AM cells lack the SOC mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5644
(±)-Methoxyverapamil hydrochloride, ≥98%
C28H40N2O5 · HCl