Acta odontologica latinoamericana : AOL
Endodontic microorganism susceptibility by direct contact test.
PMID 19177855
The aim of this study was to evaluate in vitro the duration of the antimicrobial effect of endodontic sealers by means of the Direct Contact Test. The sealers tested were: Endomethasone - Septodont, Endomethasone C-Septodont, Endion-Voco, Diaket-ESPE, Pulp Canal Sealer-SybronEndo, and AH26-Dentsply DeTrey. The endodontopathic microorganisms (MO) confronted were: Staphylococcus aureus (Sa), Candida albicans (Ca), Enterococcus faecalis (Ef), Prevotella intermedia (Pi), Porphyromonas gingivalis (Pg) and Fusobacterium nucleatum (Fn). Test specimens of each sealer were prepared and placed on the surface of agar plates that had been inoculated with each MO, and after predetermined periods, transfers were made from the contact area between the test specimen and the cultured agar and from the area that had not been in contact with the test specimens (control). The results were read as presence/absence of microbial growth and analyzed statistically using the Kruskal-Wallis test. It was concluded that the structural features and virulence of endodontopathic microorganisms determine their response to the sealers, independently of the time during which sealers act and the mechanism by which the antiseptic reaches the microorganism, which in this case was by direct contact.
Related Materials
Product #
Molecular Formula
Add to Cart
03854 Creosote from beechwood tar