EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Disulfide-linked, gold nanoparticle based reagent for detecting small molecular weight thiols.
PMID 19178279
Abstract
Disulfide-linked gold nanoparticles (AuNP) were synthesized by reacting dithiobis[succinimidylpropionate] (DSP) coated nanoparticles with glutathione disulfide. AuNP-cross-linking was monitored by the red shift and broadening of the AuNP's localized surface plasmon absorption resonance (LSPR) spectrum. The exposure of the disulfide-linked AuNPs to a variety of free thiols with systematically varying molecular weight revealed a AuNP-disulfide stability to reduction by thiols up to a critical molecular weight, M(c), of >300 Da thus making the disulfide-linked AuNP the first reagent that can discriminate thiols based on their size.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3669
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder
C14H16N2O8S2