EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Detection of cysteine conjugate metabolite adduct formation with specific mitochondrial proteins using antibodies raised against halothane metabolite adducts.
PMID 1917965
Abstract
Antibodies raised against halothane metabolite adducts cross-react with S-(1,1,2,2-tetrafluoroethyl)-L-cysteine (TFEC) and S-(2-chloro-1,1,2-trifluoroethyl)-L-cysteine metabolite adducts. Using these antibodies in immunohistochemical experiments, metabolite binding was localized to the damaged areas of the proximal tubule after treatment of male rats with TFEC. Immunoblot analysis of subcellular fractions of rat kidney tissue after in vivo treatment with TFEC revealed a high specificity for binding of metabolites to proteins of the mitochondrial fraction. These proteins may represent target molecules which play a role in cysteine conjugate induced nephrotoxicity.