EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the triterpenes from loquat leaf (Eriobotrya japonica)].
PMID 19180956
Abstract
To investigate the chemical constituents from Loquat leaf (Eriobotrya japonica). Compounds were isolated by a combination of silica gel and ODS column chromatographies and preparative TLC and HPLC, their structures were identified by chemical and spectral methods. Nine triterpenes were isolated and identified as methyl betuliate (I), oleanolic acid (II), ursolic acid (III), methyl maslinate (IV), methyl corosolate, (methyl 2alpha-3alpha-dihydroxyurs-12-en-28-oate, V), maslinic acid (VI), corosolic acid (VII), tormentic acid (VIII), euscaphic acid (IX) from loquat leaf. Compounds I and V are obtained from this plant and Eriobotrya genus for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0549
Corosolic acid from Lagerstroemia speciosa, ≥98% (HPLC)
C30H48O4