EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Deprotection of the indole (N(ind))-formyl (For) group on tryptophan employing a new reagent, N,N'-dimethylethylendiamine (DMEDA) in an aqueous solution.
PMID 19182415
Abstract
The deprotection of the indole (N(ind))-formyl (For) group on Trp was achieved in a 95% yield using N,N'-dimethylethylendiamine (DMEDA) (1.5, 2.0, 3.0 eq) in water at room temperature. A new reagent was successfully applied to the deprotection of a model peptide, H-Phe-Trp(N(ind)-For)-Lys-Tyr-OH, to give H-Phe-Trp-Lys-Tyr-OH in a 91% yield.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D157805
N,N′-Dimethylethylenediamine, 98%
C4H12N2
270032
N,N′-Dimethylethylenediamine, 85%
C4H12N2