EMAIL THIS PAGE TO A FRIEND
The FEBS journal
Crystal structures of two novel sulfonylurea herbicides in complex with Arabidopsis thaliana acetohydroxyacid synthase.
PMID 19187232
Abstract
Acetohydroxyacid synthase (AHAS; EC 2.2.1.6) is the first enzyme in the biosynthetic pathway of the branched-chain amino acids. It catalyzes the conversion of two molecules of pyruvate into 2-acetolactate or one molecule of pyruvate and one molecule of 2-ketobutyrate into 2-aceto-2-hydroxybutyrate. AHAS requires the cofactors thiamine diphosphate (ThDP), Mg(2+) and FAD for activity. The herbicides that target this enzyme are effective in protecting a broad range of crops from weed species. However, resistance in the field is now a serious problem worldwide. To address this, two new sulfonylureas, monosulfuron and monosulfuron ester, have been developed as commercial herbicides in China. These molecules differ from the traditional sulfonylureas in that the heterocyclic ring attached to the nitrogen atom of the sulfonylurea bridge is monosubstituted rather than disubstituted. The structures of these compounds in complex with the catalytic subunit of Arabidopsis thaliana AHAS have been determined to 3.0 and 2.8 A, respectively. In both complexes, these molecules are bound in the tunnel leading to the active site, such that the sole substituent of the heterocyclic ring is buried deepest and oriented towards the ThDP. Unlike the structures of Arabidopsis thaliana AHAS in complex with the classic disubstituted sulfonylureas, where ThDP is broken, this cofactor is intact and present most likely as the hydroxylethyl intermediate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34322
Chlorosulfuron, PESTANAL®, analytical standard
C12H12ClN5O4S