EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Magnesium-catalyzed asymmetric direct aldol addition of ethyl diazoacetate to aromatic, aliphatic, and alpha,beta-unsaturated aldehydes.
PMID 19191696
Abstract
Magnesium-catalyzed enantioselective aldol between ethyl diazoacetate and aromatic, aliphatic, and alpha,beta-unsaturated aldehydes affords alpha-diazo-beta-hydroxy-esters in high enantioselectivities. Aldol adducts resulting from this asymmetric transformation are versatile intermediates toward the synthesis of several ester containing chiral building blocks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E22201
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
C4H6N2O2