EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Expression of the phosphonoalanine-degradative gene cluster from Variovorax sp. Pal2 is induced by growth on phosphonoalanine and phosphonopyruvate.
PMID 19191873
Abstract
The phosphonopyruvate hydrolase (PalA) found in Variovorax sp., Pal2, is a novel carbon-phosphorus bond cleavage enzyme, which is expressed even in the presence of high levels of phosphate, thus permitting phosphonopyruvate to be used as the sole carbon and energy source. Analysis of the regions adjacent to the palA gene revealed the presence of the five structural genes that constitute the 2-amino-3-phosphonopropionic acid (phosphonoalanine)-degradative operon. Reverse transcriptase-PCR (RT-PCR) experiments demonstrated that all five genes in the operon are transcribed as a single mRNA and that their transcription is induced by phosphonoalanine or phosphonopyruvate. Transcriptional fusions of the regulatory region of the phosphonoalanine degradative operon with the gfp gene were constructed. Expression analysis indicated that the presence of a LysR-type regulator (encoded by the palR gene) is essential for the transcription of the structural genes of the operon. Similar gene clusters were found in the sequenced genomes of six bacterial species from the Alpha-, Beta- and Gammaproteobacteria, and analysis of metagenomic libraries revealed that sequences related to palA are widely spread in the marine environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4910
DL-2-Amino-3-phosphonopropionic acid
C3H8NO5P