EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Cloning, recombinant production, crystallization and preliminary X-ray diffraction analysis of a family 101 glycoside hydrolase from Streptococcus pneumoniae.
PMID 19194003
Abstract
Streptococcus pneumoniae is a serious human pathogen that is responsible for a wide range of diseases including pneumonia, meningitis, septicaemia and otitis media. The full virulence of this bacterium is reliant on carbohydrate processing and metabolism, as revealed by biochemical and genetic studies. One carbohydrate-processing enzyme is a family 101 glycoside hydrolase (SpGH101) that is responsible for catalyzing the liberation of galactosyl beta1,3-N-acetyl-D-galactosamine (Galbeta1,3GalNAc) alpha-linked to serine or threonine residues of mucin-type glycoproteins. The 124 kDa catalytic module of this enzyme (SpGH101CM) was cloned and overproduced in Escherichia coli and purified. Crystals were obtained in space group P2(1) and diffracted to 2.0 A resolution, with unit-cell parameters a = 81.86, b = 88.91, c = 88.77 A, beta = 112.46 degrees. SpGH101CM also qualitatively displayed good activity towards the synthetic substrate p-nitrophenyl-2-acetamido-2-deoxy-3-O-(beta-D-galactopyranosyl)-alpha-D-galactopyranoside, which is consistent with the classification of this enzyme as an endo-alpha-N-acetylgalactosaminidase.