EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
New thermolytic carbamoyl groups for the protection of nucleobases.
PMID 19194584
Abstract
It was found that N-arylcarbamoyl and N-(phenylsulfonyl)carbamoyl (psc) groups could be effectively introduced onto the amino groups of deoxycytidine and deoxyadenosine derivatives and could be removed thermolytically. We succeeded in synthesizing DNA probes incorporating these thermo-removable protecting groups and developed a new system for molecular switching by changing the protection- and deprotection-modes using simple heating and re-carbamoylation with isocyanates. This reversible process enabled us to control the hybridization ability of the DNA probes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09699
Ammonium carbamate, for decomposition
CH6N2O2
292834
Ammonium carbamate, 99%
CH6N2O2