EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Synthesis of dimethyl carbonate by transesterification of ethylene carbonate over activated dawsonites.
PMID 19197926
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
676802
Ethylene carbonate, anhydrous, 99%
C3H4O3
E26258
Ethylene carbonate, 98%
C3H4O3