EMAIL THIS PAGE TO A FRIEND
Psychiatry research
The Gambling Symptom Assessment Scale (G-SAS): a reliability and validity study.
PMID 19200607
Abstract
Two hundred seven patients with DSM-IV Pathological Gambling Disorder completed both the Gambling Symptom Assessment Scale (G-SAS) and the Yale-Brown Obsessive-Compulsive Scale--modified for Pathological Gambling (PG-YBOCS) at baseline visit and weekly or biweekly thereafter during the 12-week study period. The week 1 to week 2 visit data were used to assess test-retest reliability. Weekly or biweekly data were used for the G-SAS validity. The PG-YBOCS reliability and validity data have been published previously. We used the PG-YBOCS as the established scale and compared the G-SAS performance with the PG-YBOCS. Test-retest reliability was statistically significant. The correlations between the G-SAS and the PG-YBOCS and Clinical Global Impression rating were excellent. Findings suggest that the G-SAS is reliable and valid in assessing changes in symptoms during a drug treatment study.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5770
Nalmefene
C21H25NO3