EMAIL THIS PAGE TO A FRIEND
Microbiology (Reading, England)
The Streptococcus equi prophage-encoded protein SEQ2045 is a hyaluronan-specific hyaluronate lyase that is produced during equine infection.
PMID 19202092
Abstract
Streptococcus equi causes equine 'strangles'. Hyaluronate lyases, which degrade connective tissue hyaluronan and chondroitins, are thought to facilitate streptococcal invasion of the host. However, prophage-encoded hyaluronate lyases are hyaluronan-specific and are thought to be primarily involved in the degradation of the hyaluronan capsule of streptococci during bacteriophage infection. To understand the role of prophage-encoded hyaluronate lyases further, we have biochemically characterized such a hyaluronate lyase, SEQ2045 from S. equi, and have shown that it is produced during equine infection. Prophage-encoded hyaluronan-specific hyaluronate lyases may therefore play a more direct role in disease pathogenesis than previously thought.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1136
Hyaluronidase from Streptomyces hyalurolyticus, lyophilized powder