EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Correlation between the stability and redox potential of three homologous cytochromes c from two thermophiles and one mesophile.
PMID 19202290
Abstract
The stability of the oxidized and reduced forms of three homologous cytochromes c from two thermophiles and one mesophile was systematically monitored by means of Soret absorption measurements in the presence of various concentrations of a denaturant, guanidine thiocyanate, at pH 7.0 at 25 degrees C. Thermophilic Hydrogenobacter thermophilus cytochrome c(552) was the most stable in both redox states, followed by moderately thermophilic Hydrogenophilus thermoluteolus cytochrome c(552), and then mesophilic Pseudomonas aeruginosa cytochrome c(551). Further stability and electrochemical analysis of the three proteins and the reciprocal variants, which exhibited a different hydrophobic interaction with the heme, showed that the one with the higher stability in both redox states had the lower redox potential. Consequently, these cytochromes c probably adapted to the cellular environments of the original bacteria with correlated stability and redox potential constraints, which are in part regulated by the hydrophobicity around the heme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50983
Guanidine thiocyanate solution, BioUltra, for molecular biology, ~6 M in H2O
CH5N3 · CHNS