EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology and therapeutics
Pharmacokinetic study of S-1.
PMID 19203542
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
755303
3-Amino-2-fluoropropionic acid, 95%
C3H6FNO2