EMAIL THIS PAGE TO A FRIEND
Journal of Food Protection
Comparative evaluation of biofilm formation and tolerance to a chemical shock of pathogenic and nonpathogenic Escherichia coli O157:H7 strains.
PMID 19205478