EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Disposition of human drug preparations in the horse. I. Rectally administered indomethacin.
PMID 1920602
Abstract
A high-performance liquid chromatographic method to measure urinary indomethacin levels is described. In 0.5 ml urine, 1 micrograms/ml of indomethacin could be detected. Alkaline hydrolysis of urine resulted in the decomposition of indomethacin. When two suppositories of Indocid corresponding to 200 mg indomethacin were administered rectally to four horses the drug was rapidly absorbed and remained detectable in urine from 1 to 12 h. The excretion rate peaked after 2-3 h while the maximal concentration ranged from 18.5 to 80.6 micrograms/ml. Only 8 to 16% of the indomethacin dose was eliminated in urine after 12 h. A fraction of the dose was excreted as the glucuronide conjugate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5521
Suxibuzone
C24H26N2O6