EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Mecillinam: a low-risk antimicrobial agent for induction of Clostridium difficile infection in an in vitro human gut model.
PMID 19211576
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33447
Mecillinam, VETRANAL, analytical standard
C15H23N3O3S